4-(4-Chlorobenzyl)piperidine hydrochloride

Lieferant: Apollo Scientific
Achtung

OR12872-1G
APOSOR12872-1GEA 348 EUR
APOSOR12872-1G
4-(4-Chlorobenzyl)piperidine hydrochloride
4-(4-Chlorobenzyl)piperidine hydrochloride
Formel: C₁₂H₁₆ClN·ClH
Molecular Weight: 246,176 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03840138
CAS-Nummer: 36968-94-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR