4-(3,4-Dichlorobenzyl)piperidine hydrochloride

Lieferant: Apollo Scientific
Achtung

OR12873-1G
APOSOR12873-1GEA 134 EUR
APOSOR12873-1G
4-(3,4-Dichlorobenzyl)piperidine hydrochloride
4-(3,4-Dichlorobenzyl)piperidine hydrochloride
Formel: C₁₂H₁₅Cl₂N·ClH
Molecular Weight: 280,621 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03840139
CAS-Nummer: 1171138-69-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR