tert-Butyl-3-phenylpiperazine-1-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Phenylpiperazine, N1-BOC protected, 1-Boc-3-Phenylpiperazine

OR12887-250MG
APOSOR12887-250MGEA 24.8 EUR
APOSOR12887-250MG
tert-Butyl-3-phenylpiperazine-1-carboxylate
tert-Butyl-3-phenylpiperazine-1-carboxylate
Formel: C₁₅H₂₂N₂O₂
Molecular Weight: 262,35 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03840082
CAS-Nummer: 502649-25-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR