tert-Butyl-3-(4-chlorophenyl)piperazine-1-carboxylate

Lieferant: Apollo Scientific

Synonyme: 3-(4-Chlorophenyl)piperazine, N1-BOC protected, tert-Butyl 3-(4-chlorophenyl)piperazine-1-carboxylate

OR12888-1G
APOSOR12888-1GEA 190 EUR
APOSOR12888-1G
tert-Butyl-3-(4-chlorophenyl)piperazine-1-carboxylate
tert-Butyl-3-(4-chlorophenyl)piperazine-1-carboxylate
Formel: C₁₅H₂₁ClN₂O₂
Molecular Weight: 296,7924 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03840084
CAS-Nummer: 886767-49-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR