3-[(Pyridin-2-yl)oxy]benzylamine hydrochloride 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-[3-(Aminomethyl)phenoxy]pyridine hydrochloride, {3-[(Pyridin-2-yl)oxy]phenyl}methylamine hydrochloride

OR12890-100MG
APOSOR12890-100MGEA 277 EUR
APOSOR12890-100MG
3-[(Pyridin-2-yl)oxy]benzylamine hydrochloride 95%
3-[(Pyridin-2-yl)oxy]benzylamine hydrochloride
Formel: C₁₂H₁₂N₂O·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD03840096
CAS-Nummer: 1107060-73-0
UN: 1759
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR