4-[(Pyridin-2-yl)oxy]benzylamine hydrochloride
Lieferant: Apollo Scientific
|
Gefahr
|
OR12891-250MG
APOSOR12891-250MGEA
213
EUR
APOSOR12891-250MG
4-[(Pyridin-2-yl)oxy]benzylamine hydrochloride
4-[(Pyridin-2-yl)oxy]benzylamine hydrochloride
Formel:
C₁₂H₁₂N₂O·ClH Lagertemperatur: Kühlschrank |
MDL:
MFCD03840152 CAS-Nummer: 1171884-78-8 UN: 1759 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...