3-[(Pyrazin-2-yl)oxy]benzaldehyde

Lieferant: Apollo Scientific
Achtung

OR12897-100MG
APOSOR12897-100MGEA 0 EUR
APOSOR12897-100MG
3-[(Pyrazin-2-yl)oxy]benzaldehyde
3-[(Pyrazin-2-yl)oxy]benzaldehyde
Formel: C₁₁H₈N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03840111
CAS-Nummer: 887344-44-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR