4-[(Pyrazin-2-yl)oxy]benzaldehyde

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(4-Formylphenoxy)pyrazine

OR12898-100MG
APOSOR12898-100MGEA 215 EUR
APOSOR12898-100MG
4-[(Pyrazin-2-yl)oxy]benzaldehyde
4-[(Pyrazin-2-yl)oxy]benzaldehyde
Formel: C₁₁H₈N₂O₂
Molecular Weight: 200,193 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02667635
CAS-Nummer: 866156-93-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR