3-(2-Pyrimidinyloxy)benzenecarbaldehyde

Lieferant: Apollo Scientific

OR12899-1G OR12899-100MG OR12899-250MG OR12899-500MG OR12899-2.5G
APOSOR12899-1GEA 0 EUR
APOSOR12899-1G APOSOR12899-100MG APOSOR12899-250MG APOSOR12899-500MG APOSOR12899-2.5G
3-(2-Pyrimidinyloxy)benzenecarbaldehyde
3-(2-Pyrimidinyloxy)benzenecarbaldehyde
Formel: C₁₁H₈N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03791149
CAS-Nummer: 263349-26-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR