4-(Chlorsulfonyl)benzoesäure 95+%

Lieferant: Apollo Scientific

OR12901-25G
APOSOR12901-25GEA 157 EUR
APOSOR12901-25G
4-(Chlorsulfonyl)benzoesäure 95+%
4-(Chlorsulfonyl)benzoesäure
Formel: C₇H₅ClO₄S
Molecular Weight: 220,63 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00007448
CAS-Nummer: 10130-89-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR