3-[(Pyridin-3-yl)methyl]piperidine-3-carboxylic acid, N-BOC protected

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(tert-Butoxycarbonyl)-3-[(pyridin-3-yl)methyl]piperidine-3-carboxylic acid, 1-(tert-Butoxycarbonyl)-3-carboxy-3-[(pyridin-3-yl)methyl]piperidine

OR12918-100MG
APOSOR12918-100MGEA 438 EUR
APOSOR12918-100MG
3-[(Pyridin-3-yl)methyl]piperidine-3-carboxylic acid, N-BOC protected
3-[(Pyridin-3-yl)methyl]piperidine-3-carboxylic acid, N-BOC protected
Formel: C₁₇H₂₄N₂O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD03840045
CAS-Nummer: 887344-18-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR