{2-[(4-Chlorophenyl)sulphonyl]ethyl}amine hydrochloride

Lieferant: Apollo Scientific
Achtung

OR12933-100MG
APOSOR12933-100MGEA 65.4 EUR
APOSOR12933-100MG
{2-[(4-Chlorophenyl)sulphonyl]ethyl}amine hydrochloride
{2-[(4-Chlorophenyl)sulphonyl]ethyl}amine hydrochloride
Formel: C₈H₁₁Cl₂NO₂S
Molecular Weight: 256,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03840166
CAS-Nummer: 85052-88-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR