3-(4-Methylphenoxy)benzoic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Carboxy-4'-methyldiphenyl ether, 4-(3-Carboxyphenoxy)toluene

OR12938-50MG
APOSOR12938-50MGEA 42.2 EUR
APOSOR12938-50MG
3-(4-Methylphenoxy)benzoic acid
3-(4-Methylphenoxy)benzoic acid
Formel: C₁₄H₁₂O₃
Molecular Weight: 228,243 g/mol
Schmelzpunkt: 152,6 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03840106
CAS-Nummer: 62507-86-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR