6-Hydroxy-2-tetralone

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,4-Dihydro-6-hydroxynaphthalen-2(1H)-one

OR12939-250MG
APOSOR12939-250MGEA 91.5 EUR
APOSOR12939-250MG
6-Hydroxy-2-tetralone
6-Hydroxy-2-tetralone
Formel: C₁₀H₁₀O₂
Molecular Weight: 162.19 g/mol
Siedepunkt: 125 °C/3 mmHg (Subl.)
Schmelzpunkt: 140…146 °C
Lagertemperatur: Kühlschrank
MDL: MFCD05663283
CAS-Nummer: 52727-28-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR