1,2,3-Thiadiazol-4-carbonsäure
Lieferant: Apollo Scientific
|
Achtung
|
OR1294-5G
APOSOR1294-5GEA
172
EUR
APOSOR1294-5G
1,2,3-Thiadiazol-4-carbonsäure
1,2,3-Thiadiazole-4-carboxylic acid
Formel:
C₃H₂N₂O₂S Molecular Weight: 130.13 g/mol Schmelzpunkt: 227…228 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052099 CAS-Nummer: 4100-13-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...