3-(2-Methoxyphenoxy)benzoic acid

Lieferant: Apollo Scientific
Achtung

OR12941-250MG
APOSOR12941-250MGEA 174 EUR
APOSOR12941-250MG
3-(2-Methoxyphenoxy)benzoic acid
3-(2-Methoxyphenoxy)benzoic acid
Formel: C₁₄H₁₂O₄
Schmelzpunkt: 148…149 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03840108
CAS-Nummer: 500884-43-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR