2-(2-Methylphenoxy)benzoic acid 95%

Lieferant: Apollo Scientific
Achtung

OR12944-1G OR12944-5G
APOSOR12944-1GEA 0 EUR
APOSOR12944-1G APOSOR12944-5G
2-(2-Methylphenoxy)benzoic acid 95%
2-(2-Methylphenoxy)benzoic acid
Formel: C₁₄H₁₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00439674
CAS-Nummer: 6325-68-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR