2-(4-Methylphenoxy)benzoic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR12947-100MG
APOSOR12947-100MGEA
50.9
EUR
APOSOR12947-100MG
2-(4-Methylphenoxy)benzoic acid 95%
2-(4-Methylphenoxy)benzoic acid
Formel:
C₁₄H₁₂O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00439673 CAS-Nummer: 21905-69-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...