2-(2-Methoxyphenoxy)benzoic acid 97%
Lieferant: Apollo Scientific
OR12948-250MG
APOSOR12948-250MGEA
209
EUR
APOSOR12948-250MG
2-(2-Methoxyphenoxy)benzoic acid 97%
2-(2-Methoxyphenoxy)benzoic acid
Formel:
C₁₄H₁₂O₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD02220804 CAS-Nummer: 21905-73-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...