[4-(2-Methoxyphenoxy)phenyl]methylamine hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(2-Methoxyphenoxy)benzylamine hydrochloride, 2-[4-(Aminomethyl)phenoxy]anisole hydrochloride

OR12955-100MG
APOSOR12955-100MGEA 203 EUR
APOSOR12955-100MG
[4-(2-Methoxyphenoxy)phenyl]methylamine hydrochloride
[4-(2-Methoxyphenoxy)phenyl]methylamine hydrochloride
Formel: C₁₄H₁₅NO₂·ClH
Molecular Weight: 265,735 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03840155
CAS-Nummer: 1169974-82-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR