2-Amino-1-(2-methylphenyl)ethan-1-ol oxalate

Lieferant: Apollo Scientific
Achtung

OR12965-250MG
APOSOR12965-250MGEA 0 EUR
APOSOR12965-250MG
2-Amino-1-(2-methylphenyl)ethan-1-ol oxalate
2-Amino-1-(2-methylphenyl)ethan-1-ol oxalate
Formel: C₉H₁₃NO·C₂H₂O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD04117772
CAS-Nummer: 1170238-97-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR