[3-(4-Methylphenyl)-1H-pyrazol-1-yl]acetic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-[1-(Carboxymethyl)-1H-pyrazol-3-yl]toluene, 1-(Carboxymethyl)-3-(4-methylphenyl)-1H-pyrazole

OR1297-250MG
APOSOR1297-250MGEA 139 EUR
APOSOR1297-250MG
[3-(4-Methylphenyl)-1H-pyrazol-1-yl]acetic acid
[3-(4-Methylphenyl)-1H-pyrazol-1-yl]acetic acid
Formel: C₁₂H₁₂N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00665965
CAS-Nummer: 959573-30-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR