1-(2-Amino-3-methylbut-1-yl)-1H-imidazole dihydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(1H-Imidazol-1-yl)-1-isopropylethylamine dihydrochloride, 1-(1H-Imidazol-1-yl)-3-methylbutan-2-amine dihydrochloride
OR12979-100MG
APOSOR12979-100MGEA
267
EUR
APOSOR12979-100MG
1-(2-Amino-3-methylbut-1-yl)-1H-imidazole dihydrochloride
1-(2-Amino-3-methylbut-1-yl)-1H-imidazole dihydrochloride
Formel:
C₈H₁₅N₃·2ClH Molecular Weight: 226,147 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04117727 CAS-Nummer: 1172472-37-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...