3-(4-Chlorobenzyl)piperidine hydrochloride
Lieferant: Apollo Scientific
OR12985-1G
APOSOR12985-1GEA
0
EUR
APOSOR12985-1G
3-(4-Chlorobenzyl)piperidine hydrochloride
3-(4-Chlorobenzyl)piperidine hydrochloride
Formel:
C₁₂H₁₆ClN·ClH Lagertemperatur: Raumtemperatur |
MDL:
MFCD04113622 CAS-Nummer: 1171692-94-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...