3-(4-Methylbenzyl)piperidine hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-[(Piperidin-3-yl)methyl]toluene hydrochloride

OR12987-1G
APOSOR12987-1GEA 161 EUR
APOSOR12987-1G
3-(4-Methylbenzyl)piperidine hydrochloride
3-(4-Methylbenzyl)piperidine hydrochloride
Formel: C₁₃H₁₉N·ClH
Molecular Weight: 225,758 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04113625
CAS-Nummer: 625454-26-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR