2-(Aminomethyl)pyrimidine oxalate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(Pyrimidin-2-yl)methylamine ethane-1,2-dioate, 2-(Aminomethyl)-1,3-diazine oxalate
OR12993-50MG
APOSOR12993-50MGEA
49.3
EUR
APOSOR12993-50MG
2-(Aminomethyl)pyrimidine oxalate
2-(Aminomethyl)pyrimidine oxalate
Formel:
C₅H₇N₃·C₂H₂O₄ Molecular Weight: 199,164 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04117723 CAS-Nummer: 866625-10-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...