Cyclopentylmethanesulphonyl chloride

Lieferant: Apollo Scientific
Gefahr

OR12996-1G OR12996-250MG OR12996-5G
APOSOR12996-1GEA 0 EUR
APOSOR12996-1G APOSOR12996-250MG APOSOR12996-5G
Cyclopentylmethanesulphonyl chloride
Cyclopentylmethanesulphonyl chloride
Formel: C₆H₁₁ClO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD05863880
CAS-Nummer: 242459-85-4
UN: 1760
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR