3-(3-Chlorphenyl)propionsäure

Lieferant: Apollo Scientific
Gefahr

OR13022-25G
APOSOR13022-25GEA 66.7 EUR
APOSOR13022-25G
3-(3-Chlorphenyl)propionsäure
3-(3-Chlorphenyl)propionsäure
Formel: C₉H₉ClO₂
Molecular Weight: 184.62 g/mol
Schmelzpunkt: 72…76 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01310801
CAS-Nummer: 21640-48-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR