4-Hydroxy-3-methoxybenzylamin Hydrochlorid 98%

Lieferant: Apollo Scientific
Achtung

OR13027-25G OR13027-10G
APOSOR13027-25GEA 0 EUR
APOSOR13027-25G APOSOR13027-10G
4-Hydroxy-3-methoxybenzylamin Hydrochlorid 98%
4-Hydroxy-3-methoxybenzylamin Hydrochlorid
Formel: C₈H₁₂ClNO₂
Schmelzpunkt: 219…221 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012864
CAS-Nummer: 7149-10-2
EINECS: 230-468-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR