Picolinamidin Hydrochlorid

Lieferant: Apollo Scientific
Achtung

OR1303-1G OR1303-25G OR1303-5G
APOSOR1303-1GEA 0 EUR
APOSOR1303-1G APOSOR1303-25G APOSOR1303-5G
Picolinamidin Hydrochlorid
Picolinamidin Hydrochlorid
Formel: C₆H₈ClN₃
Schmelzpunkt: 145…152 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052271
CAS-Nummer: 51285-26-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR