4-Aminopyrimidine-5-carboxylic acid 98%

Lieferant: Apollo Scientific
Achtung

OR13043-1G OR13043-5G OR13043-25G
APOSOR13043-1GEA 21.9 EUR
APOSOR13043-1G APOSOR13043-5G APOSOR13043-25G
4-Aminopyrimidine-5-carboxylic acid 98%
4-Aminopyrimidine-5-carboxylic acid
Formel: C₅H₅N₃O₂
Molecular Weight: 139.11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00047383
CAS-Nummer: 20737-41-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR