2,5-Dibrom-3-picolin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,5-Dibrom-3-methylpyridin
OR13063-100G
APOSOR13063-100GEA
59.6
EUR
APOSOR13063-100G
2,5-Dibrom-3-picolin
2,5-Dibrom-3-picolin
Formel:
C₆H₅Br₂N Molecular Weight: 250.92 g/mol Schmelzpunkt: 43…47 °C Flammpunkt: >110 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02093085 CAS-Nummer: 3430-18-0 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...