5-Brom-2-ethoxypyridin 97%
Lieferant: Apollo Scientific
|
Gefahr
|
OR13065-5G
APOSOR13065-5GEA
21.9
EUR
APOSOR13065-5G
5-Brom-2-ethoxypyridin 97%
5-Brom-2-ethoxypyridin
Formel:
C₇H₈BrNO Molecular Weight: 202,05 g/mol Schmelzpunkt: 32…36 °C Flammpunkt: 103 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00234311 CAS-Nummer: 55849-30-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...