4-Iodo-3-nitrophenol

Lieferant: Apollo Scientific
Achtung

OR13076-5G
APOSOR13076-5GEA 119 EUR
APOSOR13076-5G
4-Iodo-3-nitrophenol
4-Iodo-3-nitrophenol
Formel: C₆H₄INO₃
Molecular Weight: 265,01 g/mol
Schmelzpunkt: 156 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01320680
CAS-Nummer: 113305-56-9
UN: 2811
ADR: 6.1,I

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR