Biphenylen
Lieferant: Apollo Scientific
|
Achtung
|
OR13086-100MG
OR13086-250MG
APOSOR13086-100MGEA
87
EUR
APOSOR13086-100MG
APOSOR13086-250MG
Biphenylen
Biphenylene
Formel:
C₁₂H₈ Schmelzpunkt: 113…114 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001110 CAS-Nummer: 259-79-0 UN: 3077 ADR: 9+EHS,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...