1,4-Diethinylbenzol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: p-Diethynylbenzene, 1,4-Diethylnylbenzene

OR13101-1G OR13101-5G OR13101-10G
APOSOR13101-1GEA 40.6 EUR
APOSOR13101-1G APOSOR13101-5G APOSOR13101-10G
1,4-Diethinylbenzol 97%
1,4-Diethynylbenzene
Formel: C₁₀H₆
Molecular Weight: 126.16 g/mol
Schmelzpunkt: 92 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00078375
CAS-Nummer: 935-14-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR