4-Chlor-3-nitrophenylboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
OR13107-1G
OR13107-5G
OR13107-25G
APOSOR13107-1GEA
21.9
EUR
APOSOR13107-1G
APOSOR13107-5G
APOSOR13107-25G
4-Chlor-3-nitrophenylboronsäure
4-Chlor-3-nitrophenylboronsäure
Formel:
C₆H₅BClNO₄ Schmelzpunkt: 275 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02258950 CAS-Nummer: 151169-67-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...