4-(2-Carboxyethyl)phenylboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
OR13109-1G
OR13109-5G
APOSOR13109-1GEA
94.4
EUR
APOSOR13109-1G
APOSOR13109-5G
4-(2-Carboxyethyl)phenylboronsäure
4-(2-Carboxyethyl)phenylboronsäure
Formel:
C₉H₁₁BO₄ Schmelzpunkt: 146…148 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01318119 CAS-Nummer: 166316-48-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...