3,5-Dinitro-2-methylphenylboronsäure 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,5-Dinitro-2-methylbenzeneboronic acid

OR13110-1G
APOSOR13110-1GEA 21.9 EUR
APOSOR13110-1G
3,5-Dinitro-2-methylphenylboronsäure 96%
3,5-Dinitro-2-methylphenylboronsäure
Formel: C₇H₇BN₂O₆
Molecular Weight: 225,95 g/mol
Schmelzpunkt: 122…128 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00757434
CAS-Nummer: 24341-76-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR