2-Chlor-4-ethoxyphenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chloro-4-ethoxybenzeneboronic acid

OR13119-5G
APOSOR13119-5GEA 105 EUR
APOSOR13119-5G
2-Chlor-4-ethoxyphenylboronsäure 98%
2-Chlor-4-ethoxyphenylboronsäure
Formel: C₈H₁₀BClO₃
Molecular Weight: 200.43 g/mol
Schmelzpunkt: 104…108 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02684318
CAS-Nummer: 313545-44-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR