2,2'-Bithiophen-5-carbaldehyd
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,2-Bithiophene-5-carboxaldehyde, 5-Formyl-2,2'-bithiophene
OR1312-1G
APOSOR1312-1GEA
36.4
EUR
APOSOR1312-1G
2,2'-Bithiophen-5-carbaldehyd
2,2'-Bithiophene-5-carboxaldehyde
Formel:
C₉H₆OS₂ Molecular Weight: 194,27 g/mol Siedepunkt: 210 °C Schmelzpunkt: 57 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052294 CAS-Nummer: 3779-27-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...