4-(4-Oxopiperidin-1-carbonyl)phenylboronsäure 95%

Lieferant: Apollo Scientific
Achtung

OR13121-100MG
APOSOR13121-100MGEA 21.9 EUR
APOSOR13121-100MG
4-(4-Oxopiperidin-1-carbonyl)phenylboronsäure 95%
4-(4-Oxopiperidin-1-carbonyl)phenylboronsäure
Formel: C₁₂H₁₄BNO₄
Schmelzpunkt: 156…160 °C
Lagertemperatur: Kühlschrank
MDL: MFCD04115679
CAS-Nummer: 850568-23-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR