3-(Phenylcarbamoyl)benzeneboronic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: (3-(Phenylcarbamoyl)phenyl)boronic acid, 3-[(Phenylamino)carbonyl]benzeneboronic acid, 3-Phenylaminocarbonylphenylboronic acid, 3-Borono-N-phenylbenzamide

OR13127-250MG
APOSOR13127-250MGEA 21.9 EUR
APOSOR13127-250MG
3-(Phenylcarbamoyl)benzeneboronic acid
3-(Phenylcarbamoyl)benzeneboronic acid
Formel: C₁₃H₁₂BNO₃
Schmelzpunkt: 250…260 °C
Lagertemperatur: Kühlschrank
MDL: MFCD04115711
CAS-Nummer: 397843-71-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR