Methylisonicotinate-N-oxide

Lieferant: Apollo Scientific
Achtung

OR13135-25G
APOSOR13135-25GEA 161 EUR
APOSOR13135-25G
Methylisonicotinate-N-oxide
Methylisonicotinate-N-oxide
Formel: C₇H₇NO₃
Schmelzpunkt: 121…123 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00128856
CAS-Nummer: 3783-38-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR