2-Chloro-N-(2,3-dihydro-1,4-benzodioxin-6-yl)acetamide

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chloro-N-(2,3-dihydrobenzo[b][1,4]dioxin-6-yl)acetamide

OR13149-1G
APOSOR13149-1GEA 34.8 EUR
APOSOR13149-1G
2-Chloro-N-(2,3-dihydro-1,4-benzodioxin-6-yl)acetamide
2-Chloro-N-(2,3-dihydro-1,4-benzodioxin-6-yl)acetamide
Formel: C₁₀H₁₀ClNO₃
Molecular Weight: 227,65 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00495167
CAS-Nummer: 42477-07-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR