3-(3-Phenoxyphenyl)acrylic acid

Lieferant: Apollo Scientific
Achtung

OR13152-1G OR13152-500MG
APOSOR13152-1GEA 0 EUR
APOSOR13152-1G APOSOR13152-500MG
3-(3-Phenoxyphenyl)acrylic acid
3-(3-Phenoxyphenyl)acrylic acid
Formel: C₁₅H₁₂O₃
Schmelzpunkt: 114…115 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00736661
CAS-Nummer: 77124-20-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR