3-Chlor-4-methoxyphenylboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Chloro-4-methoxybenzeneboronic acid, (3-Chloro-4-methoxyphenyl)boronic acid

OR13155-25G
APOSOR13155-25GEA 231 EUR
APOSOR13155-25G
3-Chlor-4-methoxyphenylboronsäure
3-Chlor-4-methoxyphenylboronsäure
Formel: C₇H₈BClO₃
Molecular Weight: 186.40 g/mol
Schmelzpunkt: 238…242 °C
Lagertemperatur: Kühlschrank
MDL: MFCD04039888
CAS-Nummer: 175883-60-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR