3-Chlor-2-methylphenylboronsäure 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: (3-Chloro-2-methylphenyl)boronic acid, 3-Chloro-2-methylphenyboronic acid, 3-Chloro-2-methylbenzeneboronic acid

OR13157-1G
APOSOR13157-1GEA 21.9 EUR
APOSOR13157-1G
3-Chlor-2-methylphenylboronsäure 96%
3-Chlor-2-methylphenylboronsäure
Formel: C₇H₈BClO₂
Molecular Weight: 170,40 g/mol
Schmelzpunkt: 175…176 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04115641
CAS-Nummer: 313545-20-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR