5-(4-Nitrophenyl)oxazol 95%

Lieferant: Apollo Scientific

OR1316-1G
APOSOR1316-1GEA 0 EUR
APOSOR1316-1G
5-(4-Nitrophenyl)oxazol 95%
5-(4-Nitrophenyl)oxazole
Formel: C₉H₆N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00085148
CAS-Nummer: 1014-23-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR