4-Acetamido-3-chlorbenzolsulfonylchlorid

Lieferant: Apollo Scientific
Gefahr

OR1317-250MG
APOSOR1317-250MGEA 65.4 EUR
APOSOR1317-250MG
4-Acetamido-3-chlorbenzolsulfonylchlorid
4-Acetamido-3-chlorbenzolsulfonylchlorid
Formel: C₈H₇Cl₂NO₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD00052363
CAS-Nummer: 16761-18-5
UN: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR